Merck
  • Regulation of Glucose-Dependent Golgi-Derived Microtubules by cAMP/EPAC2 Promotes Secretory Vesicle Biogenesis in Pancreatic β Cells.
Regulation of Glucose-Dependent Golgi-Derived Microtubules by cAMP/EPAC2 Promotes Secretory Vesicle Biogenesis in Pancreatic β Cells.
Current biology : CB (2019-07-16)
Kathryn P Trogden, Xiaodong Zhu, Justin S Lee, Christopher V E Wright, Guoqiang Gu, Irina Kaverina
ABSTRACT
The microtubule (MT) network is an essential regulator of insulin secretion from pancreatic β cells, which is central to blood-sugar homeostasis. We find that when glucose metabolism induces insulin secretion, it also increases formation of Golgi-derived microtubules (GDMTs), notably with the same biphasic kinetics as insulin exocytosis. Furthermore, GDMT nucleation is controlled by a glucose signal-transduction pathway through cAMP and its effector EPAC2. Preventing new GDMT nucleation dramatically affects the pipeline of insulin production, storage, and release. There is an overall reduction of β-cell insulin content, and remaining insulin becomes retained within the Golgi, likely because of stalling of insulin-granule budding. While not preventing glucose-induced insulin exocytosis, the diminished granule availability substantially blunts the amount secreted. Constant dynamic maintenance of the GDMT network is therefore critical for normal β-cell physiology. Our study demonstrates that the biogenesis of post-Golgi carriers, particularly large secretory granules, requires ongoing nucleation and replenishment of the GDMT network.
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
8-(4-Chlorophenylthio)-2′-O-methyladenosine 3′,5′-cyclic monophosphate monosodium hydrate, ≥98% (HPLC)
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
Mycophenolic acid, ≥98%