Merck
Myofibril Assembly in Cultured Mouse Neonatal Cardiomyocytes.
Anatomical record (Hoboken, N.J. : 2007) (2018-12-12)
Jennifer White, Jushuo Wang, Yingli Fan, Dipak K Dube, Joseph W Sanger, Jean M Sanger
ABSTRACT
The formation of myofibrils was analyzed in neonatal mouse cardiomyocytes grown in culture and stained with fluorescent antibodies directed against myofibrillar proteins. The cardiomyocyte cultures also were exposed to siRNA probes to test the role of nonmuscle myosin IIB expression in the formation of myofibrils. In culture, new myofibrils formed in the spreading cell margins surrounding contractile myofibrils previously assembled in utero. Observations indicated that assembly of mature myofibrils occurred in three-stages, as previously reported in cultured mouse skeletal muscle. Premyofibrils, characterized by minisarcomeres with nonmuscle myosin IIB and muscle-specific alpha-actinin bound to actin filaments, formed in the first stage; followed by nascent myofibrils, the second stage when muscle myosin II and titin were first detected. In the mature myofibril stage muscle myosin II filaments aligned in periodic A-Bands; late assembling proteins, including myomesin and telethonin, were integrated in the sarcomeres, and nonmuscle IIB was absent from the sarcomeres. Treatment of the cultured neonatal cardiomyocytes with gene-specific siRNAs for nonmuscle myosin IIB, led to a marked decrease in the formation of premyofibrils, and subsequently of mature myofibrils. Anat Rec, 301:2067-2079, 2018. © 2018 Wiley Periodicals, Inc.
Sigma-Aldrich
Monoclonal Anti-α-Actinin (Sarcomeric) antibody produced in mouse, clone EA-53, ascites fluid
Sigma-Aldrich
Anti-Myosin IIB, non-muscle antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-MYH14 antibody produced in rabbit, affinity isolated antibody