Merck
  • The JmjC domain histone demethylase Ndy1 regulates redox homeostasis and protects cells from oxidative stress.
The JmjC domain histone demethylase Ndy1 regulates redox homeostasis and protects cells from oxidative stress.
Molecular and cellular biology (2008-10-08)
Christos Polytarchou, Raymond Pfau, Maria Hatziapostolou, Philip N Tsichlis
ABSTRACT
The histone H3 demethylase Ndy1/KDM2B protects cells from replicative senescence. Changes in the metabolism of reactive oxygen species (ROS) are important for establishing senescence, suggesting that Ndy1 may play a role in redox regulation. Here we show that Ndy1 protects from H(2)O(2)-induced apoptosis and G(2)/M arrest and inhibits ROS-mediated signaling and DNA damage, while knockdown of Ndy1 has the opposite effects. Consistent with these observations, whereas Ndy1 overexpression promotes H(2)O(2) detoxification, Ndy1 knockdown inhibits it. Ndy1 promotes the expression of genes encoding the antioxidant enzymes aminoadipic semialdehyde synthase (Aass), NAD(P)H quinone oxidoreductase-1 (Nqo1), peroxiredoxin-4 (Prdx4), and serine peptidase inhibitor b1b (Serpinb1b) and represses the expression of interleukin-19. At least two of these genes (Nqo1 and Prdx4) are regulated directly by Ndy1, which binds to specific sites within their promoters and demethylates promoter-associated histone H3 dimethylated at K36 and histone H3 trimethylated at K4. Simultaneous knockdown of Aass, Nqo1, Prdx4, and Serpinb1b in Ndy1-expressing cells to levels equivalent to those detected in control cells was sufficient to suppress the Ndy1 redox phenotype.
Sigma-Aldrich
Antioxidant Assay Kit, sufficient for 200 tests
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-8-Oxoguanine Antibody, clone 483.15, ascites fluid, clone 483.15, Chemicon®