Merck
  • The daughter centriole controls ciliogenesis by regulating Neurl-4 localization at the centrosome.
The daughter centriole controls ciliogenesis by regulating Neurl-4 localization at the centrosome.
The Journal of cell biology (2017-04-08)
Abdelhalim Loukil, Kati Tormanen, Christine Sütterlin
ABSTRACT
The two centrioles of the centrosome differ in age and function. Although the mother centriole mediates most centrosome-dependent processes, the role of the daughter remains poorly understood. A recent study has implicated the daughter centriole in centriole amplification in multiciliated cells, but its contribution to primary ciliogenesis is unclear. We found that manipulations that prevent daughter centriole formation or induce its separation from the mother abolish ciliogenesis. This defect was caused by stabilization of the negative ciliogenesis regulator CP110 and was corrected by CP110 depletion. CP110 dysregulation may be caused by effects on Neurl-4, a daughter centriole-associated ubiquitin ligase cofactor, which was required for ciliogenesis. Centrosome-targeted Neurl-4 was sufficient to restore ciliogenesis in cells with manipulated daughter centrioles. Interestingly, early during ciliogenesis, Neurl-4 transiently associated with the mother centriole in a process that required mother-daughter centriole proximity. Our data support a model in which the daughter centriole promotes ciliogenesis through Neurl-4-dependent regulation of CP110 levels at the mother centriole.
Sigma-Aldrich
Anti-TTBK2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
Anti-ODF2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Tubulin Antibody, Detyrosinated, Chemicon®, from rabbit
Sigma-Aldrich
Anti-LRRC45 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Centrin Antibody, clone 20H5, clone 20H5, from mouse