Merck
  • Structure-activity study of furyl aryloxazole fluorescent probes for the detection of singlet oxygen.
Structure-activity study of furyl aryloxazole fluorescent probes for the detection of singlet oxygen.
PloS one (2018-07-03)
Renzo P Zanocco, Roger Bresoli-Obach, Santi Nonell, Else Lemp, Antonio L Zanocco
ABSTRACT
In this study, we report the synthesis and the photochemical behavior of a series of new "click-on" fluorescent probes designed to detect singlet oxygen. They include a highly fluorescent chemical structure, an aryloxazole ring, linked to a furan moiety operating as singlet oxygen trap. Their activity depends on both the structure of the aryloxazole fluorophore and the electron-donating and electron-accepting properties of the substituents attached to the C-5 of the furan ring. All probes are selectively oxidized by singlet oxygen to give a single fluorescent product in methanol and produce negligible amounts of singlet oxygen themselves by self-sensitization. The most promising dyad, (E)-2-(2-(5-methylfuran-2-yl)vinyl)naphtho[1,2-d]oxazole, FN-6, shows outstanding reactivity and sensitivity: it traps singlet oxygen with a rate constant (5,8 ± 0.1) x 107 M-1 s-1 and its fluorescence increases by a factor of 500 upon reaction. Analysis of the dyads reactivity in terms of linear free energy relationships using the modified Swain and Lupton parameter F and the Fukui condensed function for the electrophilic attack, suggests that cycloaddition of singlet oxygen to the furan ring is partially concerted and possibly involves an exciplex with a "more open" structure than could be expected for a concerted cycloaddition.
Sigma-Aldrich
2-Furoyl chloride, 95%
Sigma-Aldrich
5-Bromo-2-furaldehyde, 97%
Sigma-Aldrich
5-Phenyl-2-furaldehyde, 96%
Sigma-Aldrich
Perinaphthenone, 97%