
Zolmitriptan Related Compound E

Synonym(s):
(4S)-4-[[3-[2-(Dimethyloxidoamino)ethyl]-1H-indol-5-yl]methyl]-2-oxazolidinone, (S)-N,N-Dimethyl-2-{5-[(2-oxooxazolidin-4-yl)methyl]-1H-indol-3-yl}ethanamine oxide, Zolmitriptan N-oxide
Empirical Formula (Hill Notation):
C16H21N3O3
CAS Number:
Molecular Weight:
303.36
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2