
Vedaprofen

Synonym(s):
(±)-2-(4-Cyclohexylnaphthalen-1-yl)propionic acid
Empirical Formula (Hill Notation):
C19H22O2
CAS Number:
Molecular Weight:
282.38
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing