
Thioperamide maleate salt

Synonym(s):
MR-12842, N-Cyclohexyl-4-(1H-imidazol-4-yl)-1-piperidinecarbothioamide maleate salt
Empirical Formula (Hill Notation):
C15H24N4S · C4H4O4
CAS Number:
Molecular Weight:
408.52
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing