
Tepraloxydim

Synonym(s):
trans-2-[1-(3-Chloroallyloxyimino)propyl]-3-hydroxy-5-(tetrahydro-2H-pyran-4-yl)-2-cyclohexen-1-one
Empirical Formula (Hill Notation):
C17H24ClNO4
CAS Number:
Molecular Weight:
341.83
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2