
(S)-(−)-Styrene oxide

Synonym(s):
(S)-(−)-Phenyloxirane, (S)-Phenylethylene oxide
Empirical Formula (Hill Notation):
C8H8O
CAS Number:
Molecular Weight:
120.15
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing