
Sodium Saccharin

Synonym(s):
Saccharin sodium salt hydrate, 2,3-Dihydro-3-oxobenzisosulfonazole sodium salt, 2-Sulfobenzoic acid imide sodium salt, o-Sulfobenzimide sodium salt, Saccharin soluble
Empirical Formula (Hill Notation):
C7H4NNaO3S · xH2O
Products
Product Category
Brand
Formula Weight
Format
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing