
Sodium ionophore VI

Synonym(s):
Bis[(12-crown-4)methyl] dodecylmethylmalonate, Dodecylmethylmalonic acid bis[(12-crown-4)methyl ester]
Empirical Formula (Hill Notation):
C34H62O12
CAS Number:
Molecular Weight:
662.85
Products
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing