
Sodium acetate-2-13C

Synonym(s):
Acetic-2-13C acid sodium salt
Linear Formula:
13CH3CO2Na
CAS Number:
Molecular Weight:
83.03
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing