
Rutoside trihydrate

Synonym(s):
Rutin trihydrate, Quercetin-3-rutinoside trihydrate, Vitamin P trihydrate
Empirical Formula (Hill Notation):
C27H30O16 · 3H2O
CAS Number:
Molecular Weight:
664.56
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing