
(R)-(+)-2,2-Dimethyl-1,3-dioxolane-4-carboxaldehyde

Synonym(s):
(R)-(+)-Glyceraldehyde acetonide, (R)-Isopropylideneglyceraldehyde, NSC 89869
Empirical Formula (Hill Notation):
C6H10O3
CAS Number:
Molecular Weight:
130.14
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing