
Quercetin

Synonym(s):
Quercetin hydrate
Empirical Formula (Hill Notation):
C15H10O7 · xH2O
Products
Brand
Formula Weight
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing