
Prilocaine Related Compound A

Synonym(s):
o-Toluidine hydrochloride, 2-Methylaniline hydrochloride
Linear Formula:
CH3C6H4NH2 · HCl
CAS Number:
Molecular Weight:
143.61
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2