
Poly(N-isopropylacrylamide), amine terminated

Synonym(s):
NIPAM polymer, PNIPAM-NH2, Polyacrylamide, functionalized polyNIPAM, functionalized polyacrylamide, polyNIPAM
Linear Formula:
H(C6H11NO)nSCH2CH2NH2
Products
Brand
Physical Form
Quality Segment
Polymer Composition
Polymer Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing