
Phenoxyethanol

Synonym(s):
2-Phenoxyethanol, Ethylene glycol monophenyl ether, Phenylglycol
Linear Formula:
C6H5OCH2CH2OH
CAS Number:
Molecular Weight:
138.16
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing