
Omeprazole Related Compound I

Synonym(s):
Omeprazole sulfone N-oxide
Empirical Formula (Hill Notation):
C17H19N3O5S
Molecular Weight:
377.41
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2