
Olomoucine

Synonym(s):
2-(Hydroxyethylamino)-6-benzylamino-9-methylpurine, Olomoucine
Empirical Formula (Hill Notation):
C15H18N6O
CAS Number:
Molecular Weight:
298.34
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2