
Nutrient Mixture F-10 Ham

Synonym(s):
Ham’s F-10
Products
Product Category
Brand
Physical Form
HEPES (mM)
L-Glutamine
Sodium Bicarbonate
Phenol Red
Quality Segment
Sterilization Method
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing