Skip to Content
Merck
Norphenylephrine Hydrochloride Pharmaceutical Secondary Standard; Certified Reference Material

Norphenylephrine hydrochloride

Norphenylephrine Hydrochloride Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
α-Aminomethyl-3-hydroxybenzyl alcohol hydrochloride, 3,α-Dihydroxyphenethylamine hydrochloride, Norfenefrine hydrochloride, Norphenylephrine hydrochloride
Linear Formula:
HOC6H4CH(CH2NH2)OH · HCl
CAS Number:
Molecular Weight:
189.64
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique