Skip to Content
Merck
N,N,N′,N′-Tetrakis(2-pyridylmethyl)ethylenediamine

N,N,N′,N′-Tetrakis(2-pyridylmethyl)ethylenediamine

N,N,N′,N′-Tetrakis(2-pyridylmethyl)ethylenediamine
Synonym(s):
TPEN
Empirical Formula (Hill Notation):
C26H28N6
CAS Number:
Molecular Weight:
424.54

Products

Formula Weight
Physical Form