
N-Nitroso-N,N-bis(7-methyloctyl)amine

Empirical Formula (Hill Notation):
C18H38N2O
CAS Number:
Molecular Weight:
298.51
Products
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2