
Nitidine chloride

Synonym(s):
2,3-Dimethoxy-12-methyl-[1,3]benzodioxolo[5,6-c]phenanthridinium chloride, NSC 146397
Empirical Formula (Hill Notation):
C21H18NO4 · Cl
CAS Number:
Molecular Weight:
383.82
Products
Formula Weight
Manufacturer
Product Line
Format
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing