
N-Formyl-Met-Leu-Phe

Synonym(s):
Chemotactic peptide, N-Formyl-L-methionyl-L-leucyl-L-phenylalanine, fMLF, fMLP (misnomer but widely used)
Empirical Formula (Hill Notation):
C21H31N3O5S
CAS Number:
Molecular Weight:
437.55
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Product Line
Purity
Quality Segment
Application
Optical Isomer
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing