
Negundoside

Synonym(s):
(1S,4aS,7S,7aS)-1,4a,5,6,7,7a-Hexahydro-7-hydroxy-1-[[2-O-(4-hydroxybenzoyl)-β-D-glucopyranosyl]oxy]-7-methyl-cyclopenta[c]pyran-4-carboxylic acid, 2′-p-Hydroxybenzoyl mussaenosidic acid
Empirical Formula (Hill Notation):
C23H28O12
CAS Number:
Molecular Weight:
496.46
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing