
N-Acetylbenzocaine

Synonym(s):
Ethyl 4-acetamidobenzoate
Empirical Formula (Hill Notation):
C11H13NO3
CAS Number:
Molecular Weight:
207.23
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing