Skip to Content
Merck
mono-tert-Butyl malonate 95%

mono-tert-Butyl malonate

mono-tert-Butyl malonate 95%
Linear Formula:
(CH3)3COCOCH2COOH
CAS Number:
Molecular Weight:
160.17

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing