
Monoclonal Anti-DNP−Alkaline Phosphatase Conjugate antibody produced in mouse

Synonym(s):
Anti-Dinitrophenyl, Monoclonal Anti-DNP
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Conjugate
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing