
Monoclonal Anti-c-Myc−FITC antibody produced in mouse

Synonym(s):
Anti-c-Myc
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing