
Metoprolol EP Impurity F

Synonym(s):
1-[(1-Methylethyl)amino]-3-phenoxy-2-propanol
Empirical Formula (Hill Notation):
C12H19NO2
CAS Number:
Molecular Weight:
209.28
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing