![Methyl parahydroxybenzoate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/100/935/4e9463bf-7abe-4633-8d19-6196865c92fb/640/4e9463bf-7abe-4633-8d19-6196865c92fb.png)
Methyl parahydroxybenzoate
![Methyl parahydroxybenzoate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/100/935/4e9463bf-7abe-4633-8d19-6196865c92fb/640/4e9463bf-7abe-4633-8d19-6196865c92fb.png)
Synonym(s):
Methyl 4-hydroxybenzoate, p-Hydroxybenzoic acid methyl ester, Methyl paraben, NIPAGIN
Linear Formula:
HOC6H4CO2CH3
CAS Number:
Molecular Weight:
152.15
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing