
Madecassoside

Synonym(s):
Asiaticoside A
Empirical Formula (Hill Notation):
C48H78O20
CAS Number:
Molecular Weight:
975.12
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing