
Loxapine succinate

Synonym(s):
Loxapine succinate salt
Empirical Formula (Hill Notation):
C18H18ClN3O · C4H6O4
CAS Number:
Molecular Weight:
445.90
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing