
Losartan Impurity G

Synonym(s):
Triphenylmethanol, Triphenylcarbinol, Trityl alcohol
Linear Formula:
(C6H5)3COH
CAS Number:
Molecular Weight:
260.33
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing