Skip to Content
BY
EN
Back
L-Linalool
Synonym(s):
(−)-Linalool, (
R
)-(−)-3,7-Dimethyl-1,6-octadien-3-ol
Linear Formula:
(CH
3
)
2
C=CHCH
2
CH
2
C(CH
3
)(OH)CH=CH
2
CAS Number:
126-91-0
Molecular Weight:
154.25
EC Number:
204-811-2
Products
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
W263516
L-Linalool,
natural, ≥95%, FG
Expand
View Pricing