![Levocarnitine Related Compound A pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/379/545/6c37839f-e21d-4262-80df-70f5ed27c418/640/6c37839f-e21d-4262-80df-70f5ed27c418.jpg)
Levocarnitine Related Compound A
![Levocarnitine Related Compound A pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/379/545/6c37839f-e21d-4262-80df-70f5ed27c418/640/6c37839f-e21d-4262-80df-70f5ed27c418.jpg)
Synonym(s):
3-Carboxy-N,N,N-trimethyl-2-propen-1-aminium chloride, Crotonobetaine hydrochloride, Crotonoylbetaine hydrochloride
Empirical Formula (Hill Notation):
C7H14ClNO2
CAS Number:
Molecular Weight:
179.64
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2