
Indole-5-carbonitrile

Synonym(s):
5-Cyanoindole
Empirical Formula (Hill Notation):
C9H6N2
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing