
Hydroxyvalerenic acid

Synonym(s):
Valerenolic acid
Empirical Formula (Hill Notation):
C15H22O3
CAS Number:
Molecular Weight:
250.33
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing