Skip to Content
Merck
Guanosine 5′-diphosphate tris salt from Saccharomyces cerevisiae Type VI, ≥92.5%

Guanosine 5′-diphosphate tris salt from Saccharomyces cerevisiae

Guanosine 5′-diphosphate tris salt from Saccharomyces cerevisiae Type VI, ≥92.5%
Synonym(s):
GDP
Empirical Formula (Hill Notation):
C10H15N5O11P2
CAS Number:
Molecular Weight:
443.20

Products

Brand
Formula Weight
Purity
Quality Segment
Showing 1-1 of 1