
Fmoc-Cys(Trt)-OH

Synonym(s):
Fmoc-Cys(Trt)-OH, N-(9-Fluorenylmethoxycarbonyl)-S-trityl-L-cysteine, Nα-Fmoc-S-trityl-L-cysteine
Empirical Formula (Hill Notation):
C37H31NO4S
CAS Number:
Molecular Weight:
585.71
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing