
Enterolactone

Synonym(s):
trans-α,β-Bis(3-hydroxybenzyl)butyrolactone
Empirical Formula (Hill Notation):
C18H18O4
CAS Number:
Molecular Weight:
298.33
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing