
DL-3-Hydroxynorvaline

Synonym(s):
α-Amino-β-hydroxyvaleric acid, 2-Amino-3-hydroxypentanoic acid, 3-Hydroxy-2-aminopentanoic acid, DL-β-Hydroxynorvaline
Empirical Formula (Hill Notation):
C5H11NO3
CAS Number:
Molecular Weight:
133.15
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application
Amino Acid
Showing 1-1 of 1
Compare
Product Number
Description
Pricing