(Dimethylamino)trimethyltin(IV)
Synonym(s):
Pentamethylstannanamine
Linear Formula:
(CH3)3SnN(CH3)2
CAS Number:
Molecular Weight:
207.89
Products
Brand
Boiling Point (Ā°C)
Formula Weight
Physical Form
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing