Skip to Content
Merck
Diazepam certified reference material, pharmaceutical secondary standard

Diazepam

Diazepam certified reference material, pharmaceutical secondary standard
Synonym(s):
7-Chloro-1-methyl-5-phenyl-3H-1,4-benzodiazepin-2(1H)-one, Ro 5-2807
Empirical Formula (Hill Notation):
C16H13ClN2O
CAS Number:
Molecular Weight:
284.74

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade