Skip to Content
Merck
Cystamine dihydrochloride purum, ≥98.0% (AT)

Cystamine dihydrochloride

Cystamine dihydrochloride purum, ≥98.0% (AT)
Synonym(s):
2,2′-Diaminodiethyl disulfide dihydrochloride, 2,2′-Dithiobis(ethylamine) dihydrochloride, Decarboxycystine dihydrochloride
Linear Formula:
NH2CH2CH2SSCH2CH2NH2 · 2HCl
CAS Number:
Molecular Weight:
225.20
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Special Grade