
Cromolyn sodium

Synonym(s):
Cromolyn sodium salt, Cromoglycate, Cromoglycic acid
Linear Formula:
C23H14O11Na2
CAS Number:
Molecular Weight:
512.33
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade