
Corning® Transwell® 44 cm2 , cell growth area, 13 mL , 100 mm , dish, 9 mL , 75 mm plate inserts

Products
Product Category
Manufacturer
Binding Type
Diameter (mm)
Surface Area cm²
Working Volume (ml)
Color
Membrane Material
Package Size
Pore size (µm)
Sterility
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing